D4898-16
p-NITROANILINE
This test method covers the determination of insoluble contamination in hydraulic fluids by gravimetric analyses. The contamination determined includes both particulate and gel-like matter, organic and inorganic, which is retained on a membrane filter disk of pore diameter as required by applicable specifications (usually 0.45 μm or 0.80 μm).
Analysis Type (Application Method): Gravimetric
Link to ASTM® D4898-16 Method
Compatible Filters:
Vacuum glassware plus accessories. Forceps to handle filter membrane.