Chromolith® WP 300 RP-18, RP-8 and RP-4: Reversed-Phase HPLC Columns for bio-applications
Reversed phase chromatography is often used for protein and peptide separations. The longer octadecyl (C18) chains, can efficiently separate complex peptide mixtures; shorter C8 modified columns are used for small, lesshydrophobic proteins; C4 is mainly applied for separation of hydrophobic proteins.
Separation of five peptides on a Chromolith® WP 300 RP-18, 100-4.6mm
Separation of five peptides on a Chromolith® WP 300 RP-18, 100-4.6mm
Separation of peptide/protein mixture on a Chromolith® WP 300 RP-8, 100-4.6mm
Separation of peptide/protein mixture on a Chromolith® WP 300 RP-8, 100-4.6mm
Analysis of Cetuximab® sample on a Chromolith® WP 300 RP-4, 100-4.6mm
Analysis of Cetuximab® sample on a Chromolith® WP 300 RP-4, 100-4.6mm
Materials Table
Product Modification I.d. Length Type Content Product No.
Chromolith® WP 300 Protein A Protein A 4.6 mm 25 mm   1 HPLC column 152258
Chromolith® WP 300 RP18 RP-18 4.6 mm 100 mm   1 HPLC column 152270
Chromolith® WP 300 RP18 RP-18 4.6 mm 50 mm   1 HPLC column 152271
Chromolith® WP 300 RP18 Guard Cartridge RP-18 4.6 mm 10 mm b 3 guard cartridges 152272
Chromolith® WP 300 RP18 Guard Cartridge RP-18 4.6 mm 5 mm b 3 guard cartridges 152273
Chromolith® WP 300 RP8 RP-8 4.6 mm 100 mm   1 HPLC column 152265
Chromolith® WP 300 RP8 RP-8 4.6 mm 50 mm   1 HPLC column 152266
Chromolith® WP 300 RP8 Guard Cartridge RP-8 4.6 mm 10 mm b 3 guard cartridges 152267
Chromolith® WP 300 RP8 Guard Cartridge RP-8 4.6 mm 5 mm b 3 guard cartridges 152268
Chromolith® WP 300 RP4 RP-4 4.6 mm 100 mm   1 HPLC column 152260
Chromolith® WP 300 RP4 RP-4 4.6 mm 50 mm   1 HPLC column 152261
Chromolith® WP 300 RP4 Guard Cartridge RP-4 4.6 mm 10 mm b 3 guard cartridges 152262
Chromolith® WP 300 RP4 Guard Cartridge RP-4 4.6 mm 5 mm b 3 guard cartridges 152263
Chromolith® WP 300 Epoxy Epoxy 4.6 mm 100 mm   1 HPLC column 152250
Chromolith® WP 300 Epoxy Epoxy 4.6 mm 50 mm   1 HPLC column 152251
Chromolith® WP 300 Epoxy Epoxy 4.6 mm 25 mm   1 HPLC column 152252
Chromolith® WP 300 Epoxy Guard Cartridge Epoxy 4.6 mm 10 mm b 3 guard cartridges 152253
Chromolith® WP 300 Epoxy Guard Cartridge Epoxy 4.6 mm 5 mm b 3 guard cartridges 152254