Method and Software Updates - Photometer
Be Up-to-Date and Download Now!
Method Updates – For Free!
By updating your photometers, you can use the same instrument to conduct the new tests without having to program it yourself. This gives you the assurance that your results will be correct, as it’s our responsibility to calibrate the tests for the instrument. Benefit from always having an up-to-date instrument!
Download the software update easily at the click of a mouse. The use of this update is completely free of charge.
Please choose between the .exe file or the compressed .zip file. You must decompress the .zip file after you download it. Double-click the .exe file and it will unzip itself. To update the software please check the manual of the respective instrument.
Update for Spectroquant® Prove 100, 300 and 600
Prove Software Version 1.5.1
Download ZIP file
This update includes:
  • Over 20 new methods for Food & Beverage and Biology
  • Update of some existing methods
  • New features
  • Some minor bug fixes
For update instructions, please consult the operating manual (section 9.2.8).
Requirement for the update:
  • PC with Microsoft Windows: NT, 2000, XP, Vista or higher
  • A free USB port on the PC
  • Empty USB stick (no formatted data)
Update for Spectroquant® Move 100
Software update for Move 100: Version xxx.71
Download ZIP file
The following articles and program are required for updating the Colorimeter:
  • Update cable, Product No. 1.73634.0001
  • Adapter RS232 to USB
Update for Spectroquant® Photometers Nova 30 A, Nova 60 and 60 A
For all Nova photometers Method Version: 38.31
Download ZIP file
Nova Firmware Update for Nova photometers from Serial number 14230751 on, please use the following software update version 4.03. For earlier serial numbers, there is no update possible.
Download ZIP file
The following articles and programs are required for updating the photometers via the incorporated RS 232 interface:
  • PC (Windows 95 or later)
  • Nova 30 A and Nova 60 photometer
  • PC cable, Product No. 1.14667.0001
Update for Spectroquant® Spectrophotometers Pharo 100 and Pharo 300
(discontinued)
Pharo Software Version 253_250
Download ZIP file
Requirement for the update:
  • PC with Microsoft Windows: NT, 2000, XP, Vista or higher
  • A free USB port on the PC
  • Empty USB stick (formatted no data)
Update for Spectroquant® Multy Colorimeter
(discontinued)
Software Version: V012.004.3.003.071
Download ZIP file
Requirements for running the update:
  • PC running a Windows operating system
  • Data-transfer cable supplied with the unit
  • Supplied screwdriver
  • Program Hexaload.exe (see CD in operating manual)
  • Software update (=*zip file, which can be downloaded here)
Materials