Spectroquant® Prove for Complete Beer Analysis
Join our on-demand webinar on Beer analysis without the headache: Chemical and Microbiological monitoring for breweries
Focus on Brewing Quality Beer
The Spectroquant® Prove family sets the standard for the brewery industry, offering a comprehensive range of methods for analysis throughout the beer brewing process. Our brewery manual provides you with a complete set of technical methods for beer analysis – all the way from raw materials to the finished product. 
You can rely on the high quality analytical data provided by the Spectroquant® Prove system—leaving you to focus on brewing tastier beer.
Analysis Methods for the Brewery Industry Using the Spectroquant® Prove
Test methods are pre-programmed in our Spectroquant® Prove instruments for the analysis of beer and wort according to MEBAK, EBC and ASBC. The manual includes test methods for:
  • Bitterness
  • Flavonoids
  • Free Amino Nitrogen
  • Total Carbohydrates
  • Iso-α Acids
  • Total Polyphenols
  • Reducing Sugars in Malt – Henry method
  • Iron Content
These and more detailed test procedures are described in the manual, where we have compiled step-by-step guidance from reagent preparation and sample processing through to analysis. To further support your analytical success, information about test solution stability and storage that can’t be found in the standard MEBAK method descriptions have also been included.
Spectroquant® Test Kits for Increased Efficiency & Accuracy
To optimize your beer production analysis, try these Spectroquant® test kits:
Need to test sulfite levels in your beer? Check out our fast and reliable SO2 quantification method.