Spectroquant® Test Kits for the METTLER TOLEDO UV5 Spectrophotometer
You have a METTLER TOLEDO Test Tube Holder for round cells?
Request Your Discount*
For further information about using Spectroquant® for photometric analysis in
your lab.
Request More Information
Ready-to-use Spectroquant® Test Kits can be used seamlessly
with your METTLER TOLEDO spectrophotometer.
We provide a comprehensive portfolio of analytical test kits and reference standards for your water analysis. Spectroquant® Test Kits are available in two different formats:
Spectroquant® Reagent Test Kits contain all reagents for your photometric analyses and can be used with any existing cuvettes of various pathlengths, e.g. 10 mm rectangular.
Spectroquant® Cell Test Kits are ready-to-use 16 mm round cells pre-filled with reagents and no further cuvette is needed.
METTLER TOLEDO is a leading global manufacturer of precision instruments including spectrophotometers. Spectroquant® photometric consumables, from the comprehensive Supelco® portfolio of analytical products, can now be used with these instruments.
To enable the seamless use of our Spectroquant® Cell Tests, METTLER TOLEDO launched the Test Tube Holder for round cells and developed the preprogrammed methods for the below listed Spectroquant® Test Kits, which can be downloaded and imported into METTLER TOLEDO spectrophotometers. This new “Test Tube Holder” for the UV5 Spectrophotometer can be purchased at METTLER TOLEDO.
Achieve accurate and reliable results with chemical consumables of certified quality with the full range of Spectroquant® Test Kits. The following 20 cell tests can be selected by OneClick™ in the LabX photometer control software. For Spectroquant® Test Kit parameters which currently do not have a pre-programmed setting, calibration will be required.
Combine convenience with precise water analysis and use Spectroquant® Cell Test Kits with the UV5 Spectrophotometer.
Get 20% off your Spectroquant® Test Kits* with purchase of the “Test Tube Holder” from METTLER TOLEDO.
Request Your Discount*
Spectroquant® Test Kits
Preprogrammed Spectroquant® Test Kits:
Catalog No. Spectroquant® Cell
Test Parameter
Measuring range
Method ID in
UV5
Application / Segment
114544 Ammonium 0.5 – 16.0 mg/L NH4-N M9907 Wastewater
114559 Ammonium 4.0 – 80.0 mg/L NH4-N M9919 Wastewater
114739 Ammonium 0.010 – 2.000 mg/L NH4-N M9908 Wastewater, drinking water,
process water
100597 Chlorine 0.03 – 6.00 mg/L Cl2 M9909 Wastewater, drinking water,
process water
114540 COD 10 – 150 mg/L COD M9901 Wastewater
114541 COD 25 – 1500 mg/L COD M9911 Wastewater
114555 COD 500 – 10000 mg/L COD M9917 Wastewater
114560 COD 4.0 – 40.0 mg/L COD M9912 Wastewater
114542 Nitrate 0.5 – 18.0 mg/L NO3-N M9905 Wastewater, drinking water,
114563 Nitrate 0.5 – 25.0 mg/L NO3-N M9906 Wastewater, drinking water
114764 Nitrate 1.0 – 50.0 mg/L NO3-N M9914 Wastewater, drinking water
114547 Nitrite 0.010 - 0.700 mg/L NO2-N M9910 Wastewater, drinking water,
process water
114537 Nitrogen (total) 0.5 – 15.0 mg/L N M9902 Wastewater
114763 Nitrogen (total) 10 – 150 mg/L N M9913 Wastewater
114543 Phosphate 0.05 – 5.00 mg/L PO4-P M9918 Wastewater, drinking water,
process water
114546 Phosphate 0.5 – 25.0  mg/L PO4-P M9903 Process water
114729 Phosphate  0.5 – 25.0 mg/L PO4-P    M9904 Wastewater 
114564 Sulfate 100 – 1000 mg/L SO42- M9920 Wastewater, drinking water
114548 Sulfate 5 – 250 mg/L SO42- M9915 Drinking water
100961 Total Hardness 5 – 215 mg/L Ca2+ M9916 Drinking water
*Conditions for Discount:
Get 20% off list price for Spectroquant® consumables with the promotion code provided in the Test Tube Holder box. After applying your code in the request form above, you will be contacted by our sales representative.
The discount is
• for single use only
• cannot be combined with other discounts
• is applicable only in countries where METTLER TOLEDO’s Test Tube Holder is available
• applicable only for direct purchase (conditions of dealers may differ)