Testing Food for Fipronil Contamination
Background
Chick and eggsIn June 2017, eggs contaminated with the pesticide Fipronil were discovered in Belgium and the Netherlands. A treatment against ticks containing fipronil had been incorrectly used to treat poultry and, if consumed, fipronil can be harmful to humans.
The EU’s executive alarm system, the Rapid Alert System for Food and Feeds (RASFF), alerted several European countries that eggs contaminated with Fipronil may have crossed their borders, both eggs and poultry meat meant for human consumption have been affected by this scandal.
What is Fipronil?
Fipronil is an insecticide used against pests including ants, fleas, lice, ticks, cockroaches and also red mites which are commonly found in poultry. It has been shown to be acutely toxic in animal experiments when ingested orally, absorbed through the skin, or when inhaled, and should therefore not be used on food producing animals.
The maximum allowed fipronil residue levels (the sum of both fipronil and its sulfone metabolite) for chicken eggs or chicken meat is 0.005 mg/kg of body weight.
How to Accurately Test for Fipronil and Fipronil Metabolites
Samples are best prepared using the QuEChERS method followed by testing using GC/MS, GC/MS/MS or LC/MS/MS techniques. We offer a complete range of analytical standards, certified reference materials, solvents, and columns for analysis of your food samples. Additionaly we also develop application methods for determining fipronil and its sulfone metabolite in raw eggs, chicken meat and mayonnaise.
Reference Materials and Standards
Our high quality Pestanal® analytical standards and TraceCERT® certified reference materials can help ensure that measurements lower than the maximum allowed residue level can be detected, and our double accredited manufacturing process to ISO/IEC 17025 and ISO 17034 gives you the assurance of accuracy.
EFSA report: results of follow-up fipronil monitoring
Following the detection of fipronil residues in eggs due to the use of non-approved veterinary medicinal products in poultry farms against red mites, an ad-hoc monitoring program was set up in the EU. Member States provided results for over 5000 samples of eggs and chicken muscle/fat, which were analyzed for the presence of fipronil and a number of additional active substances proposed by the European Commission.
The report summarizes the data sampled during the period from 1 September 2017 to 30 November 2017 and reported to the European Food Safety Authority (EFSA). It provides an overview of the results with regards to the frequency of occurrence and compliance with the legal limits. The results are presented by country of origin, type of food product, and by the active substances analyzed. The EFSA also lists 54 pesticides, mostly acaricides, which should also be considered when analyzing egg or egg-containing samples.
To view the recommended substances by EFSA please go to table EFSA recommended pesticide standards for fipronil testing.
To view the report, click on the link under Additional information.
Materials
Product No. Description Add to Cart
16785-50MG Fipronil, certified reference material, TraceCERT®
46451-100MG Fipronil, Pestanal, analytical standard
79157-5MG Fipronil-(pyrazole-13C3, cyano-13C), analytical standard
32333-50MG Fipronil sulfone, Pestanal, analytical standard
34519-10MG Fipronil carboxamide, Pestanal, analytical standard
34520-10MG Fipronil sulfide, Pestanal, analytical standard
41865-25MG Fipronil-desulfinyl, Pestanal, analytical standard
00334-5mg Fipronil-(pyrazole-13C3, cyano-13C)sulfone, analytical standard
ERMBB125 Egg powder (fipronil) Matrix Material
Product No. Description Add to Cart
1.00029 Acetonitrile hypergrade Click on Product No. for
quantities and pricing
1.06007 Methanol gradient grade for liquid chromatography Click on Product No. for
quantities and pricing
1.06035 Methanol hypergrade for LC-MS Click on Product No. for
quantities and pricing
1.11727 Ethanol gradient grade for liquid chromatography Click on Product No. for
quantities and pricing
1.15333 Water for chromatography (LC-MS) Click on Product No. for
quantities and pricing
5.33001 Acetic acid 100% for LC-MS LiChropur Click on Product No. for
quantities and pricing
5.33002 Formic acid 98-100% for LC-MS LiChropur Click on Product No. for
quantities and pricing
5.43841 Potassium dihydrogen phosphate anhydrous for HPLC LiChropur Click on Product No. for
quantities and pricing
73594 Ammonium acetate for mass spectrometry, eluent additive for LC-MS Click on Product No. for
quantities and pricing
70221 Ammonium formate for mass spectrometry, ≥99.0% Click on Product No. for
quantities and pricing
69372 Triethylammonium acetate buffer for HPLC, 0.98-1.02M Click on Product No. for
quantities and pricing
28471-U GC-MS colums, SLB-5ms, 30m x 0.25mm I.D., 0.25μm
Product No. Description Quality Cart No.
31732-100mg Abamectin PESTANAL®, analytical standard
45331-250mg Azamethiphos PESTANAL®, analytical standard
45332-250mg Azinphos-ethyl PESTANAL®, analytical standard
74116-50mg Azinphos-ethyl certified reference material, TraceCERT®
45333-250mg Azinphos-methyl PESTANAL®, analytical standard
45336-250mg Bendiocarb PESTANAL®, analytical standard
45357-250mg Bromopropylate PESTANAL®, analytical standard
32055-250mg Carbaryl PESTANAL®, analytical standard
36856-10ml Carbaryl solution 100mg/ml in cyclohexane, PESTANAL®, analytical standard
32056-250mg Carbofuran PESTANAL®, analytical standard
37913-100mg-R Chlorfenapyr PESTANAL®, analytical standard
36124-100mg Chlorfenson PESTANAL®, analytical standard
45376-50mg Chlorobenzilate PESTANAL®, analytical standard
45395-100mg Chlorpyrifos PESTANAL®, analytical standard
94114-100mg Chlorpyrifos certified reference material, TraceCERT®
31553-2ml,
31553-10ml
Chlorpyrifos solution 100mg/ml in acetonitrile, PESTANAL®, analytical standard
CRM48104,
ampule of 1ml
Chlorpyrifos solution 1000mg/ml in methyl tert-butyl ether, certified reference material, TraceCERT®
45396-250mg Chlorpyrifos-methyl PESTANAL®, analytical standard
45831-2ml,
45831-10ml
Chlorpyrifos-methyl solution 100mg/ml in acetonitrile, PESTANAL®, analytical standard
45403-250mg Coumaphos PESTANAL®, analytical standard
46003-250mg b-Cyfluthrin PESTANAL®, analytical standard
33738-250mg Baythroid (Cyfluthrin) Mixture of isomers, PESTANAL®, analytical standard
74767-100mg Cyhalothrin PESTANAL®, analytical standard
45423-250mg Deltamethrin PESTANAL®, analytical standard
05995-50mg Deltamethrin certified reference material, TraceCERT®
45441-250mg Dichlorvos PESTANAL®, analytical standard
46391-100mg Dicyclanil PESTANAL®, analytical standard
45446-250mg Diflubenzuron PESTANAL®, analytical standard
77070-50mg Diflubenzuron certified reference material, TraceCERT®
45449-100mg Dimethoate PESTANAL®, analytical standard
33993-100mg-R Doramectin VETRANAL®, analytical standard
31733-250mg Emamectin benzoate PESTANAL®, analytical standard
32015-250mg Endosulfan Mixture of isomers, PESTANAL®, analytical standard
45852-2ml Endosulfan solution 100mg/ml in hexane, PESTANAL®, analytical standard
45468-100mg α-Endosulfan PESTANAL®, analytical standard
74119-50mg α-Endosulfan certified reference material, TraceCERT®
36659-2ml-R α-Endosulfan solution 100mg/ml  in hexane, PESTANAL®, analytical standard
33385-100mg β-Endosulfan PESTANAL®, analytical standard
40828-50mg β-Endosulfan certified reference material, TraceCERT®
36582-2ml β-Endosulfan solution 100mg/ml in hexane, PESTANAL®, analytical standard
32526-100mg Eprinomectin PESTANAL®, analytical standard
34111-2ml, 34111-10ml Eprinomectin solution 100mg/ml in acetonitrile, PESTANAL®, analytical standard
45477-250mg Ethion PESTANAL®, analytical standard
45481-250mg Etrimfos PESTANAL®, analytical standard
45487-250mg Fenitrothion PESTANAL®, analytical standard
31223-250mg Fenpropathrin PESTANAL®, analytical standard
36552-250mg Fenthion PESTANAL®, analytical standard
45495-250mg Fenvalerate PESTANAL®, analytical standard
90664-50mg Fenvalerate certified reference material, TraceCERT®
46113-100mg-R Fluazuron PESTANAL®, analytical standard
46417-100mg Flumethrin PESTANAL®, analytical standard
46424-250mg Formothion solution ~80% in xylene, PESTANAL®, analytical standard
37902-100mg-R Hexaflumuron PESTANAL®, analytical standard
31662-100mg Lufenuron PESTANAL®, analytical standard
36143-100mg Malathion PESTANAL®, analytical standard
31558-2ml,
31558-10ml
Malathion solution 100mg/ml in cyclohexane, PESTANAL®, analytical standard
91481-50mg Malathion certified reference material, TraceCERT®
33395-100mg Methamidophos PESTANAL®, analytical standard
46342-2ml Methamidophos solution 100mg/ml in acetonitrile, PESTANAL®, analytical standard
89933-50mg Methamidophos certified reference material, TraceCERT®
36158-100mg Methidathion PESTANAL®, analytical standard
74189-50mg Methidathion certified reference material, TraceCERT®
33767-2ml ( E )-Mevinphos solution 100mg/ml in acetonitrile, PESTANAL®, analytical standard
33765-2ml (Z)-Mevinphos solution 100mg/ml in acetonitrile, PESTANAL®, analytical standard
33746-25mg Moxidectin Vetranal, analytical standard
36733-1g (-) - Nicotine PESTANAL®, analytical standard
46343-10ml (-) – Nicotine solution 100mg/ml in acetonitrile, PESTANAL®, analytical standard
45607-100mg Parathion PESTANAL®, analytical standard
45890-2ml Parathion solution 100mg/ml in cyclohexane, PESTANAL®, analytical standard
36187-100mg Parathion-methyl PESTANAL®, analytical standard
45891-2ml-R Parathion-methyl solution 100mg/ml in cyclohexane, PESTANAL®, analytical standard
45614-250mg Permethrin Mixture of cis and trans isomers, PESTANAL®, analytical standard
45893-2ml-R Permethrin (isomers) solution 100mg/ml in acetonitrile, PESTANAL®, analytical standard
45622-100mg Phosphamidon PESTANAL®, analytical standard
45644-250mg Propoxur PESTANAL®, analytical standard
45664-100mg Sulfotep PESTANAL®, analytical standard
45756-250mg-R Teflubenzuron PESTANAL®, analytical standard
45680-250mg Tetradifon PESTANAL®, analytical standard
45696-50mg Triazophos PESTANAL®, analytical standard
68298-50mg Triazophos certified reference material, TraceCERT®