Two-Component Reagents for Volumetric Karl Fischer Titration
Aquastar® Two Component Reagents
In two-component reagent systems, the titrant contains only iodine and methanol, while the solvent contains the other Karl Fischer reaction components – sulfur dioxide and a suitable base dissolved in methanol. It is used as the working medium in the Karl Fischer titration cell.
  • Titration rate slightly faster
  • High accuracy
  • Better buffer capacity
  • Distinct endpoint to ensure confidence in results
  • Good titer stability
  • Ensured constant and consistent high quality
Aquastar® Titrant
Aquastar® Titrant reagents for two-component volumetric Karl Fischer analysis and contain iodine and methanol. Titrant reagents need to be combined with Aquastar Solvent.
Aquastar® Solvent
Aquastar® Solvent contains sulfur dioxide and the base; both are dissolved in alcohol. This medium is filled into the titration cell to dissolve the sample.
If you have trouble finding the products you're looking for, please contact our customer support for assistance.