Aquastar® Sodium Tartrate Dihydrate 15.66%
Sodium tartrate dihydrate is the volumetric standard for Karl Fischer titration. Under normal conditions, it is stable and non-hygroscopic. Sodium tartrate dihydrate has a stoichiometric water content of 15.66%. And, it is primarily used for titer determination in volumetry.
Materials
If you have trouble finding the products you're looking for, please contact our customer support for assistance.