Aquastar® Water Standard Oil 15–30 ppm
This standard is especially designed for users who need to measure the water content in oil samples. The water standard exactly corresponds to the low water content and the matrix of oil samples and the exact value for each batch is stated in the certificate. Practical ampoules ensure ease of handling.
If you have trouble finding the products you're looking for, please contact our customer support for assistance.