Lopinavir Assay Following European Pharmacopoeia 10 Guidelines Using an Ascentis Express C18 Column and UV Detection
Anita Piper, Scientist Instrumental Analytics R&D
Introduction
This paper illustrates how it is possible to set-up the assay method for Lopinavir testing following the current European pharmacopeia guidelines (10.2). The monograph assay method calls for a column with l = 0.25 m, Ø = 4.6 mm end-capped octadecylsilyl silica gel for chromatography with 4 µm particle size. No particular HPLC column is referenced in the EP knowledge database for assay method, and the method is of isocratic nature.
This gives a chance to replace the monograph column geometry/particle size with a shorter and faster alternative column (up to 70% reduction in length) packed with smaller particles (up to 50% reduction). This can save valuable time, and at the same time you can benefit from improved separation efficiency, which typically translates into better method performance and sensitivity. In this study, the limit of detection (LOD) is better than 1 ppm using HPLC-UV detection.
Lopinavir
Lopinavir
Experimental Conditions
Column Ascentis® Express C18 (2.7µm) 150x4.6 mm Injection: 12 µL
Detection UV = 215 nm (micro flow cell; 1.4 µL/7mm) Flow Rate: 1.0 mL/min
Mobile phase A Acetonitrile/phosphate buffer solution 45/55 (v/v) Temperature: 50 °C
Phosphate buffer Dissolve 0.9 g of dipotassium hydrogen phosphate and 2.7 g of potassium dihydrogen phosphate in 900 mL of water and mix well. Adjust to pH 6.0 with phosphoric acid, dilute to 1000 mL with water and filter. Pressure Drop: 153 bar (2219 psi)
Solvent mixture Acetonitrile/water 50/50 (v/v)    
Test solution (a) Dissolve 50.0 mg of the substance to be examines in the solvent mixture and dilute to 100 mL with the solvent mixture.
Test solution (b) Dilute 5.0 mL of the test solution (a) to 100 mL with the solvent mixture.
Reference solution (a) Dissolve 50.0 mg of Lopinavir CRS in the solvent mixture and dilute to 100 mL with the solvent mixture.
Dilute 5 mL of this solution to 100 mL with the solvent mixture.
Reference Solution (a)
Reference Solution (a)
Test Solution (b)
Test Solution (b)
No. Compound Retention Time (min) Tailing Factor
1 t0 void volume 1.1  
2 Lopinavir CRS 16.2 0.97
1. Specificity:
Inject reference solution (a) and determine the retention time and the content of desired analyte
    Retention Time (min) Area (%) Tailing Factor
1 Lopinavir CRS 16.3 96.3 0.95
2. Standard Repeatability (25 ppm)
Sample Mean Area (mAU*min)
STD 1 14.79
STD 2 14.82
STD 3 14.86
STD 4 14.90
STD 5 14.93
Mean 14.86
Standard Deviation 0.06
(%) RSD 0.4
3. LOD & LOQ
Conc. (ppm) Mean Area (mAU*min)
0.25 0.46
0.63 0.67
1.25 1.06
3.13 2.10
6.25 4.02
12.50 7.44
18.75 11.24
25.00 14.86
37.50 22.54
STEYEX 0.1348
Slope 0.5888
LOD (ppm) 0.8
LOQ (ppm) 2.3
Concentration
Concentration
Conclusion
In this study in reference to European Pharmacopeia 10 Guidelines, a shorter and faster Fused-Core® (Superficially Porous Particle, SPP) column was evaluated for an assay method for Lopinavir, achieving a limit of detection (LOD) of better than 1 ppm using HPLC-UV detection.
Materials