NIOSH 0500 (Issue 2)
PARTICULATES NOT OTHERWISE REGULATED, TOTAL
This test method uses a gravimetric (filter weight) method to monitor airborne particulate material. This method is nonspecific and determines the total dust concentration to which a worker is exposed. It may be applied, e.g., to gravimetric determination of fibrous glass in addition to the other ACGIH particulates not otherwise regulated.
Analysis Type (Application Method): Gravimetric
Link to NIOSH 0500 (Issue 2) Method
Compatible Filters:
37 mm 2 - 5 μm PVC or equivalent hydrophobic filter and supporting pad in 37 mm cassette filter holder. Forceps to handle filter membrane.