NIOSH 0600
PARTICULATES NOT OTHERWISE REGULATED, RESPIRABLE
The method measures the mass concentration of any non-volatile respirable dust. In addition to inert dusts, the method has been recommended for respirable coal dust. The method is biased in light of the recently adopted international definition of respirable dust, e.g., ≈ +7% bias for non-diesel, coal mine dust.
Analysis Type (Application Method): Gravimetric
Link to NIOSH 0600 Method
Compatible Filters:
37 mm 5 μm PVC hydrophobic membrane supported by a cassette filter holder. Forceps to handle filter membrane.