NIOSH 2542 (Issue 1)
MERCAPTANS, METHYL-, ETHYL-, and n-BUTYL-
This test method uses gas chromatography and FPD sulfur mode techniques to detect methyl, ethyl, and n-butyl mercaptans. The analytes are highly flammable and irritating to the eyes. Work in a hood. This is a modification of OSHA Method 26 for methyl mercaptan with the addition of ethyl- and n-butyl mercaptan.
Analysis Type (Application Method): GC, FPD sulfur mode
Link to NIOSH 2542 (Issue 1) Method
Compatible Filters:
37 mm glass fiber w/acrylic binder in 5% (w/v) aqueous solution of mercuric acetate, in 2-piece cassette w/o backup pad impregnated with mercuric acetate. Forceps to handle filter membrane.