NIOSH 5000
CARBON BLACK
The method uses a gravimetric (filter weight) technique for an analysis that is simple but the method is nonspecific that may be extended to high air concentrations (e.g., nuisance dust levels) by collecting a smaller sample volume. This is Method S262 in a revised format.
Analysis Type (Application Method): Gravimetric
Link to NIOSH 5000 Method
Compatible Filters:
37 mm 5 μm PVC and stainless steel support screen in cassette filter holder. Forceps to handle filter membrane.