NIOSH 5002 (Issue 2)
WARFARIN
This method uses HPLC and UV detection. Use gloves, goggles and other appropriate equipment to prevent eye contact and repeated or prolonged skin contact. This is P&CAM 313 in a new format. Warfarin is commonly used to treat blood clots.
Analysis Type (Application Method): HPLC, UV detection
Link to NIOSH 5002 (Issue 2) Method
Compatible Filters:
37 mm 1 μm PTFE and cellulose backup pad in two-piece filter holder held together with tape or shrinkable band. Forceps to handle filter membrane.