NIOSH 5004
HYDROQUINONE
This test method uses HPLC (UV detection) techniques to monitor hydroquinone. This method can be used when significant concentrations of hydroquinone vapor are not present. This is Method S57 in a revised format.
Analysis Type (Application Method): HPLC-UV
Link to NIOSH 5004 Method
Compatible Filters:
37 mm 0.8 μm MCE and a cellulose backup. Forceps to handle filter membrane.