NIOSH 5005 (Issue 2)
THIRAM
This method uses an HPLC (UV detection) technique. NIOSH researchers have used this method at facilities that use Thiram as an insecticide. Take preauction that Acetonitrile is toxic and flammable; work with it only in a hood. This is Method S256 in a revised format.
Analysis Type (Application Method): HPLC UV detection
Link to NIOSH 5005 (Issue 2) Method
Compatible Filters:
37 mm 1 μm PTFE in two piece polystyrene cassette filter holder with backup pad, sealed with tape or a shrinkable band. Forceps to handle filter membrane.