NIOSH 5007 (Issue 2)
ROTENONE
This test method uses HPLC (UV detection) techniques to detect rotenone. The method is applicable to commercial formulations. Avoid breathing acetonitrile vapors; may cause skin irritation. This is Method S300 in a revised format. Rotenone is an colorless crystalline that is used as an insecticide and pesticide.
Analysis Type (Application Method): HPLC, UV detection
Link to NIOSH 5007 (Issue 2) Method
Compatible Filters:
37 mm 1 μm PTFE with backup pad in two-piece cassette. Forceps to handle filter membrane.