NIOSH 5009
BENZOYL PEROXIDE
This method uses HPLC and UV detection. Take precaution that ethyl ether is highly flammable and forms explosive peroxides on exposure to air. This is Method S253 in a revised format. Benzoyl Peroxide is a medication that helps treat skin issue like acne. It also can be used to bleach flour, hair, whiten teeth, and bleach textiles.
Analysis Type (Application Method): HPLC-UV
Link to NIOSH 5009 Method
Compatible Filters:
37 mm 0.8 μm MCE in 37 mm filter holder. Forceps to handle filter membrane.