NIOSH 5010
(1) BROMOXYNIL and (2) BROMOXYNIL OCTANOATE
This method uses HPLC and UV detection to permit the simultaneous determination of both analytes. Bromoxynil is classified as a nitrile herbicide. A white solid, it works by inhibiting photosynthesis. Be aware it is moderately toxic to mammal and cronic exposure can cause long-term sickness.
Analysis Type (Application Method): HPLC-UV
Link to NIOSH 5010 Method
Compatible Filters:
37 mm glass fiber or 2 μm PTFE membrane, with opaque cassette filter holder. Forceps to handle filter membrane.