NIOSH 5011
ETHYLENE THIOUREA
Ethylene thiourea is used in the manufacture of fungicides, rubber vulcanization and in dyes, pharmaceuticals, synthetic resins and electroplating baths. The method was also applied to tetramethyl thiourea.
Analysis Type (Application Method): Gas chromatography, flame photometric
Link to NIOSH 5011 Method
Compatible Filters:
37 mm 5 μm PVC filter or 0.8 μm MCE in a plastic filter cassette. Forceps to handle filter membrane.