NIOSH 5012
EPN
This test method uses gas chromatography and flame photometric techniques to monitor EPN. Malathion and Parathion have also been determined by this procedure. This method replaces Method S285. The method of Hill and Arnold has also been used for analysis of pesticides. Method 5600 is an alternative method for organophosphorus pesticides. Isooctane is flammable. Prepare all samples in a well-ventilated hood.
Analysis Type (Application Method): Gas chromatography, flame photometric
Link to NIOSH 5012 Method
Compatible Filters:
37 mm glass fiber filter type AE in a 2 piece polystyrene cassette with backup pad. Forceps to handle filter membrane.