NIOSH 5014
CHLORINATED TERPHENYL (60% CHLORINE)
This test method uses gas chromatography and 63Ni ECD techniques to monitor chlorinated terphenyl. Chlorinated terphenyls may be similar to chlorinated biphenyls in toxicity. Use protective clothing to avoid skin contact.
Analysis Type (Application Method): Gas chromatography 63Ni ECD
Link to NIOSH 5014 Method
Compatible Filters:
37 mm 1 μm glass fiber supported by backup pad in polystyrene cassette. Forceps to handle filter membrane.