NIOSH 5016 (Issue 2)
STRYCHNINE
This test method is a HPLC (UV detection) technique. This revises Method S302. Note that Strychnine is a potent convulsant. Take precaution by wearing gloves, face shield, and impervious clothing when handling it.
Analysis Type (Application Method): HPLC, UV detection
Link to NIOSH 5016 (Issue 2) Method
Compatible Filters:
37 mm glass fiber filter in cassette filter holder. Syringe filter, polypropylene housing, 0.2 μm nylon. Forceps to handle filter membrane.