NIOSH 5017
DIBUTYL PHOSPHATE
[Missing from NIOSH′s page] This test method uses gas chromatography and phosphorus FPD techniques to monitor dibutyl trimethylsilyl phosphate. This methodd revises P&CAM 297. Sample preparations should be conducted in a hood.
Analysis Type (Application Method): Gas chromatography FPD
Link to NIOSH 5017 Method
Compatible Filters:
37 mm 1 μm PTFE in a 2 piece polystyrene cassette filter holder. Forceps to handle filter membrane.