NIOSH 5019
AZELAIC ACID
Azelaic acid is an organic compound with the formula HOOC(CH2)7COOH. This saturated dicarboxylic acid exists as a white powder. This method uses gas chromatography and FID and may be applicable to other straight-chain aliphatic dicarboxylic acids. Work must be performed in a fume hood. This revision replaces Method 7400, Revision #3 (dated 5/15/89).
Analysis Type (Application Method): GC, FID
Link to NIOSH 5019 Method
Compatible Filters:
5 μm PVC membrane in cassette filter holder. Forceps to handle filter membrane.