NIOSH 5022
Sulfuric Acid in Workplace Atmospheres
This test method uses an ion chromatopgrahy and hydride atomic absorption technique to monitor anions (IC); AsH3 (AAS). The method is designed to quantitate particulate organo-arsenic compounds. Arsenic Organo describes any organic compound containing a carbon to arsenic bond.
Analysis Type (Application Method): IC/hydride atomic absorption
Link to NIOSH 5022 Method
Compatible Filters:
37 mm 1 μm PTFE polyethylene-backed membrane filter w/backup pad in cassette filter holderForceps to handle filter membrane.