NIOSH 5025
CHLORINATED DIPHENYL OXIDE
This test method uses gas chromatography, electrolytic, and conductivity detection to monitor chlorinated diphenyl oxide. The method has been evaluated only for the hexachloro derivative. The sampler may not be adequate for the monochloro or dichloro derivatives, which have higher vap or pressures. This revises Method S119.
Analysis Type (Application Method): Gas chromatography, electrolytic conductivity detector
Link to NIOSH 5025 Method
Compatible Filters:
37 mm 0.8 μm MCE supported by a screen in a cassette holder. Forceps to handle filter membrane.