NIOSH 5026 (Issue 2)
OIL MIST, MINERAL
This method uses infrared spectrophotometry to monitor mineral oil. This method is applicable to all trichlorofluoroethane-soluble mineral oil mists, but not to (nor does OSHA’s standard cover) semi-synthetic or synthetic cutting fluids. This revises P&CAM 283. P&CAM 159 and S272 use similar samplers with measurement by fluorescence spectrophotometry. Carbon tetrachloride is a suspected human carcinogen. Handle in a fume hood.
Analysis Type (Application Method): IR
Link to NIOSH 5026 (Issue 2) Method
Compatible Filters:
37 mm 0.8 μm MCE, 5 μm PVC, 2 μm PTFE, or glass fiber filter, with cellulose backup pad in two-piece filter cassette. Forceps to handle filter membrane.