NIOSH 5027 (Issue 2)
RIBAVIRIN
This test method uses HPLC (UV detection) techniques to detect ribavirin. Use protective gloves when handling. Work only in a fume hood. This method is a modification of a bulk assay procedure developed by Eastman Kodak Company.
Analysis Type (Application Method): HPLC, UV detection
Link to NIOSH 5027 (Issue 2) Method
Compatible Filters:
37 mm 1 μm glass fiber filter with a cellulose backup pad in a 2-piece cassette. Forceps to handle filter membrane.