NIOSH 5029 (Issue 1)
DEMETON
The method is specific for MDA and the use of UV and electrochemical detectors in serie s gives greater reliability to the analysis, and allows a wide measurement range. The method has been applied to wipe samples.
Analysis Type (Application Method): HPLC, UV and electrochemical
Link to NIOSH 5029 (Issue 1) Method
Compatible Filters:
37 mm, acid-treated glass fiber and 0.45 μm PTFE. Forceps to handle filter membrane.