NIOSH 5030
CYANURIC ACID
Cyanuric acid can be produced by hydrolysis of crude or waste melamine followed by crystallization. It is also used as a chlorine stabilizer in swimming pools, binding to free chlorine and releases it slowly, extending the time needed to deplete each dose of sanitizer. This test method uses HPLC (UV detection) to monitor cyanuric acid. HPLC method for measurement in solution is a variation of the method of Briggle et al. Ingestion of methanol may cause blindness or death. Methanol is a fire hazard (flash point 12 °C).
Analysis Type (Application Method): HPLC-UV
Link to NIOSH 5030 Method
Compatible Filters:
37 mm 5 μm PVC membrane and 25 mm 0.45 μm PTFE in polypropylene housing. Forceps to handle filter membrane.