NIOSH 5033 (Issue 1)
p-NITROANILINE
This test method uses a HPLC (UV detection) technique to detect p-nitroaniline. This method is applicable to p-nitroaniline aerosol but not for environments where vapor may be present. This revises Method S7.
Analysis Type (Application Method): HPLC, UV detection
Link to NIOSH 5033 (Issue 1) Method
Compatible Filters:
37 mm 0.8 μm MCE and cellulose backup pad (37 mm) in a 2-piece filter holder held together by shrinkable band. Forceps to handle filter membrane.