NIOSH 5036 (Issue 1)
TRIMELLITIC ANHYDRIDE
This method uses gas chromatopgraphy and FID techniques to monitor trimellitic anhydride. Trimellitic anhydride chloride is a chemical compound used to produce polyamide-imide plastic.The method was used for analysis of bulk samples and air (field samples). This method will not differentiate between trimellitic anhydride and trimellitic acid. This revises Method P&CAM 322. Boron trifluoride and pyridine are toxic. Work with these substances only in a hood.
Analysis Type (Application Method): GC-FID
Link to NIOSH 5036 (Issue 1) Method
Compatible Filters:
37 mm 0.8 μm PVC copolymer filter supported by a cellulose backup pad in a three-piece plastic filter holder. Forceps to handle filter membrane.