NIOSH 5037 (Issue 1)
TRIORTHOCRESYL PHOSPHATE
This method uses the GC, FPD in phosphorus mode technique to adapt to other phosphates of relatively low volatility with appropriate changes in chromatographic conditions. Avoid contact of triorthocresyl phosphate with eyes, skin, and clothing. Handle only in a fume hood. This revises Method S209.
Analysis Type (Application Method): GC, FPD phosphorus mode
Link to NIOSH 5037 (Issue 1) Method
Compatible Filters:
37 mm 0.8 μm MCE supported by cellulose backup pad in three-piece filter holder. Forceps to handle filter membrane.