NIOSH 5038 (Issue 1)
TRIPHENYL PHOSPHATE
This method uses gas chromatography and FPD in phosphorous mode to adapt to other phosphates of relatively low volatility with appropriate changes in chromatographic conditions. Handle only in a fume hood. Avoid skin contact. This method revises S210.
Analysis Type (Application Method): GC, FPD phosphorus mode
Link to NIOSH 5038 (Issue 1) Method
Compatible Filters:
37 mm 0.8 μm cellulose ester with cellulose backup pad held in a three-piece cassette filter holder supported by a cellulose backup pad. Forceps to handle filter membrane.