NIOSH 5044
ESTROGENIC COMPOUNDS
This test method uses a HPLC (UV detection) technique to detect estrogenic compounds. The method is based on a preliminary investigation analyzing field wipe samples collected during a health hazard evaluation at a facility producing birth control pills. Because estrogenic compounds may be carcinogens, handle in a well ventilated hood and wear protective clothing.
Analysis Type (Application Method): HPLC
Link to NIOSH 5044 Method
Compatible Filters:
37 mm 2 μm PTFE with cellulose space ring in a 2 piece cassette filter holder