NIOSH 5504 (Issue 2)
ORGANOTIN COMPOUNDS (as Sn)
The method was validated using TeBT, TBTC, TCHH, and BuIOMA as surrogates for the most important classes of organotin compounds. If speciation of organotin compounds is not required and if inorganic tin compounds are absent, the HPLC separation may be deleted. Use special care to avoid losses of TeBT and other volatile tetra-substituted organotin compounds
Analysis Type (Application Method): HPLC, Atomic Absorption, graphite furnace
Link to NIOSH 5504 (Issue 2) Method
Compatible Filters:
37 mm glass fiber filter in a 2 piece polystyrene cassette w/ backup holder. Example compatible would be MilliporeSigma′s glass filber without binder membrane. Forceps to handle filter membrane.