NIOSH 5508
KEPONE
This test method uses gas chromatography and ECD techniques to monitor kepone. Personal and area samples were collected by this method at a Mirex formulation plant. This method was originally designated P&CAM 225. Kepone is a colorless solid and is a pollutant that is banned globally.
Analysis Type (Application Method): Gas chromatography, ECD
Link to NIOSH 5508 Method
Compatible Filters:
37 mm 0.8 μm MCE in a filter holder. Forceps to handle filter membrane.