NIOSH 5519
ENDRIN
This test method uses gas chromatography and 63Ni ECD techniques to monitor endrin. Smaller concentrations can be determined if desorption efficiency is satisfactory. This is a modification of the method of Hill and Arnold and it replaces NIOSH method S284. Handle all chemicals and organic solvents in the laboratory hood.
Analysis Type (Application Method): Gas chromatography 63Ni ECD
Link to NIOSH 5519 Method
Compatible Filters:
37 mm 0.8 μm MCE supported by a stainless steel screen in cassette filter holder. A Solid sorbent is also used in conjunction with filter. Forceps to handle filter membrane.