NIOSH 6006
DIBORANE
This test method uses plasma emission spectrometry techniques to monitor boron. This revises P&CAM 341. The measurement can also be done by inductively-coupled plasma atomic emission spectrometry (e.g., Method 7300). Diborne is a colorless gas that mixes well with air and can form explosive mixtures. Diborne can be used as a reagent in organic synthesis.
Analysis Type (Application Method): Plasma emission spectrometry
Link to NIOSH 6006 Method
Compatible Filters:
37 mm 1 μm PTFE in plastic cassette. Forceps to handle filter membrane.
Materials
Related Links
Categories