NIOSH 6007 (Issue 2)
NICKEL CARBONYL
This test method uses an atomic absorption and graphic furnace technique to detect nickel. Particulate nickel compounds will give a positive interference unless a prefilter is used during sampling. This revises P&CAM 344.
Analysis Type (Application Method): Atomic Absorption, graphite furnace
Link to NIOSH 6007 (Issue 2) Method
Compatible Filters:
37 mm 0.8 μm cellulose ester filter and cellulose backup pad in a plastic filter holder. Forceps to handle filter membrane.