NIOSH 6011 Chlorine
BROMINE
This test method uses ion chromatography and conductivity to monitor chlorine and bromide ions. This method uses ion chromatography and conductivity and has sufficient sensitivity for STEL samples. Take precaution to handle in a fume hood. P&CAM 209 (colorimetric), OSHA Methods ID-101 and ID-108 are alternative methods. Wear protective clothing. Handle in a fume hood.
Analysis Type (Application Method): Ion chromatography, conductivity
Link to NIOSH 6011 Chlorine Method
Compatible Filters:
25 mm 0.45 μm silver filter with porous plastic support pad; prefilter, 0.5 μm PTFE with PTFE support, 0.5-μm or 0.4-μm polyester. Forceps to handle filter membrane.