NIOSH 6015
AMMONIA
The purpose of this method is for the preparation and analysis of air samples for detection and measurement of ammonia. This method uses visible absorption spectrophotometry, applicable to STEL measurements. Note special precaution to avoid contact with skin or inhalation of vapors. Handle in a hood and avoid skin contact.
Analysis Type (Application Method): Prefiltration, air sampling, visible absorption spectrophotometry
Link to NIOSH 6015 Method
Compatible Filters:
Prefilter (to remove particulate interferences): 37 mm 0.8 μm cellulose ester membrane supported by stainless steel screen in 2 piece cassette filter holder. Forceps to handle filter membrane.