NIOSH 7020
CALCIUM and compounds, as Ca
This method uses Atomic Absorption (flame) to perform elemental analysis, not compound specific to verify that the types of compounds in the samples are soluble with this ashing procedure. with this method, it is possible to identify an additional four metals. Note special precaution to perform all acidic digestions in the chemical fume hood. NIOSH 7300 is an alternate analytical method.
Analysis Type (Application Method): Atomic absorption
Link to NIOSH 7020 Method
Compatible Filters:
37 mm 0.8 μm MCE in 37 mm filter holder. Forceps to handle filter membrane.