NIOSH 7024
CARBARYL
This test method uses atomic absorption and flame technqiues to monitor chromium. This is an elemental analysis for total Cr, not compound specific. Perform all acid digestions in a fume hood. This method combines and replaces P&CAM 173, P&CAM 152, S323, and S352. Method 7300 (Elements by ICP-AES) is an alternate analytical method.
Analysis Type (Application Method): Atomic absorption
Link to NIOSH 7024 Method
Compatible Filters:
37 mm 0.8 μm MCE in cassette. Forceps to handle filter membrane.