NIOSH 7027
CHROMIUM and compounds, as Cr
This test method uses atomic absorption and flame techniques to monitor cobalt. This is an elemental analysis, not compound specific. This method combines and replaces P&CAM 173 and S203 for cobalt. Method 7300 (Elements by ICP-AES) is an alternate analytical technique. Perform all acid digestions in a fume hood.
Analysis Type (Application Method): Atomic absorption
Link to NIOSH 7027 Method
Compatible Filters:
37 mm 0.8 μm MCE in a cassette filter holder. Forceps to handle filter membrane.