NIOSH 7056
BARIUM, soluble compounds
This method is designed to monitor Barium salt of nitric acid. This method uses atomic absorption (flame) to determine Ba 2+ in water-soluble barium compounds. Insoluble barium compounds (e.g.,BaSO4 ) require an ashing procedure. Wear protective clothing and work in a fume head. This revises Method S198.
Analysis Type (Application Method): Atomic absorption
Link to NIOSH 7056 Method
Compatible Filters:
37 mm 0.8 MCE in cassette filter holder. Forceps to handle filter membrane.