NIOSH 7082
LEAD by Flame AAS
The method is applicable to elemental lead, including Pb fume, and all other aerosols containing lead. This is an elemental analysis, not compound specific. Aliquots of the samples can be analyzed separately for additional elements.
Analysis Type (Application Method): Atomic absorption spectrophotometry (AAS)
Link to NIOSH 7082 Method
Compatible Filters:
37 mm 0.8 μm MCE in a cassette filter holder. Forceps to handle filter membrane.