NIOSH 7105
LEAD by GFAAS
The method is applicable to elemental lead, including Pb fume, and all other aerosols containing lead. This is an elemental analysis, not compound specific. Aliquots of the sample may be analyzed separately for additional elements.
Analysis Type (Application Method): X-Ray powder diffraction
Link to NIOSH 7105 Method
Compatible Filters:
37 mm 0.8 μm MCE in a 2 piece cassette filter holder. Forceps to handle filter membrane.